1-(2-Chloro-3-fluoropyridin-4-yl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

110896-1G 110896-500MG 110896-5G 110896-25G
77463-966EA 415.09 USD
77463-966 77476-766 77486-828 77497-682
1-(2-Chloro-3-fluoropyridin-4-yl)cyclohexanol ≥97%
1-(2-Chloro-3-fluoropyridin-4-yl)cyclohexanol

Precaución: For research use only

Formula: C₁₁H₁₃ClFNO
Storage Temperature: Ambient
MDL Number: MFCD34780869
CAS Number: 2918936-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR