3-Bromo-2-(cyclopropylmethoxy)pyridine ≥95%

Supplier: AOBChem USA

Warning

110840-1G 110840-5G 110840-25G
77463-962EA 64.89 USD
77463-962 77486-822 77497-676
3-Bromo-2-(cyclopropylmethoxy)pyridine ≥95%
3-Bromo-2-(cyclopropylmethoxy)pyridine

Precaución: For research use only

Formula: C₉H₁₀BrNO
MW: 228.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14651785
CAS Number: 1249861-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR