(4-Bromo-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥95%

Supplier: AOBChem USA

Warning

110673-1G 110673-500MG 110673-5G 110673-25G
77463-958EA 415.09 USD
77463-958 77476-760 77486-818 77497-672
(4-Bromo-2-fluoro-1,3-phenylene)bis(methylsulfane) ≥95%
(4-Bromo-2-fluoro-1,3-phenylene)bis(methylsulfane)

Precaución: For research use only

Formula: C₈H₈BrFS₂
MW: 267.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780746
CAS Number: 2918935-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR