1-Ethoxy-4-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

110262-1G 110262-500MG 110262-5G 110262-25G
77463-954EA 415.09 USD
77463-954 77476-756 77486-814 77497-666
1-Ethoxy-4-fluoro-2-methoxybenzene ≥97%
1-Ethoxy-4-fluoro-2-methoxybenzene

Precaución: For research use only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34780498
CAS Number: 2901066-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR