2-Ethoxy-6-iodophenol ≥97%

Supplier: AOBChem USA

Warning

109753-1G 109753-500MG 109753-5G 109753-25G
77463-950EA 415.09 USD
77463-950 77476-752 77486-810 77497-660
2-Ethoxy-6-iodophenol ≥97%
2-Ethoxy-6-iodophenol

Precaución: For research use only

Formula: C₈H₉IO₂
Storage Temperature: Ambient
MDL Number: MFCD34821783
CAS Number: 2639571-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR