3-Ethoxy-2-hydroxy-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Ethoxy-2-hydroxybenzoic acid

109746-1G 109746-500MG 109746-5G 109746-25G
77463-946EA 202.91 USD
77463-946 77476-748 77486-806 77497-654
3-Ethoxy-2-hydroxy-benzoic acid ≥97%
3-Ethoxy-2-hydroxy-benzoic acid

Precaución: For research use only

Formula: C₉H₁₀O₄
MW: 182.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01412200
CAS Number: 67127-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR