2-(3-Chloro-2-(methylthio)phenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

109558-1G 109558-500MG 109558-5G 109558-25G
77463-942EA 415.09 USD
77463-942 77476-744 77486-802 77497-650
2-(3-Chloro-2-(methylthio)phenyl)propan-2-ol ≥97%
2-(3-Chloro-2-(methylthio)phenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₃ClOS
Storage Temperature: Ambient
MDL Number: MFCD34821766
CAS Number: 2918967-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR