3-Bromo-5-iodo-2-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

109229-1G 109229-500MG 109229-5G 109229-25G
77463-938EA 266.77 USD
77463-938 77476-740 77486-798 77497-640
3-Bromo-5-iodo-2-methoxypyridine ≥95%
3-Bromo-5-iodo-2-methoxypyridine

Precaución: For research use only

Formula: C₆H₅BrINO
MW: 313.918 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27922996
CAS Number: 1823944-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR