2-(3-Bromo-4-chlorophenyl)-2-methyl-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

10888-1G 10888-500MG 10888-5G 10888-25G
77463-934EA 103 USD
77463-934 77476-736 77486-794 77497-636
2-(3-Bromo-4-chlorophenyl)-2-methyl-1,3-dioxolane ≥95%
2-(3-Bromo-4-chlorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619355
CAS Number: 2221812-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR