1-Bromo-2-(dimethoxymethyl)-3,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

108767-1G 108767-500MG 108767-5G 108767-25G
77463-930EA 415.09 USD
77463-930 77476-734 77486-790 77497-632
1-Bromo-2-(dimethoxymethyl)-3,4-difluorobenzene ≥97%
1-Bromo-2-(dimethoxymethyl)-3,4-difluorobenzene

Precaución: For research use only

Formula: C₉H₉BrF₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34779998
CAS Number: 2901099-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR