2,6-Bis(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Bis(trifluoromethyl)benzeneboronic acid

10864-1G 10864-5G 10864-25G
77463-926EA 71.07 USD
77463-926 77486-784 77497-626
2,6-Bis(trifluoromethyl)phenylboronic acid ≥97%
2,6-Bis(trifluoromethyl)phenylboronic acid

Precaución: For research use only

Formula: C₈H₅BF₆O₂
MW: 257.93
Storage Temperature: Ambient
MDL Number: MFCD04039321
CAS Number: 681812-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR