3-(Ethylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

108010-1G 108010-500MG 108010-5G
77463-922EA 469.68 USD
77463-922 77476-728 77486-780
3-(Ethylthio)benzaldehyde ≥97%
3-(Ethylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₁₀OS
MW: 166.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21760494
CAS Number: 87425-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR