4-Bromo-2-hydroxy-6-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

107660-1G 107660-500MG 107660-5G 107660-25G
77463-918EA 166.86 USD
77463-918 77476-724 77486-776 77497-620
4-Bromo-2-hydroxy-6-methylbenzaldehyde ≥97%
4-Bromo-2-hydroxy-6-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₇BrO₂
MW: 215.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705518
CAS Number: 1427438-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR