3-((4-Fluorobenzyl)oxy)-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

107289-1G 107289-500MG 107289-5G 107289-25G
77463-914EA 415.09 USD
77463-914 77476-720 77486-772 77497-612
3-((4-Fluorobenzyl)oxy)-5-methylbenzaldehyde ≥95%
3-((4-Fluorobenzyl)oxy)-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₃FO₂
MW: 244.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34779396
CAS Number: 1645954-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR