3-((2-Fluorobenzyl)oxy)-5-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

107282-1G 107282-500MG 107282-5G 107282-25G
77463-910EA 415.09 USD
77463-910 77476-716 77486-768 77497-608
3-((2-Fluorobenzyl)oxy)-5-methylbenzaldehyde ≥95%
3-((2-Fluorobenzyl)oxy)-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34779389
CAS Number: 2918869-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR