4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

107240-1G 107240-500MG 107240-5G
77463-906EA 811.64 USD
77463-906 77476-712 77486-764
4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde ≥95%
4-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde

Precaución: For research use only

Formula: C₈H₃BrF₄O
MW: 271.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32878885
CAS Number: 2091678-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR