Methyl 4-formyl-2-(trifluoroMethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-formyl-2-(trifluoromethyl)benzoate

107208-1G 107208-500MG 107208-5G
77463-902EA 558.26 USD
77463-902 77476-708 77486-760
Methyl 4-formyl-2-(trifluoroMethyl)benzoate ≥97%
Methyl 4-formyl-2-(trifluoroMethyl)benzoate

Precaución: For research use only

Formula: C₁₀H₇F₃O₃
MW: 232.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27935000
CAS Number: 959632-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR