1-isobutyl-4-(methoxymethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

107197-1G 107197-500MG 107197-5G 107197-25G
77463-898EA 415.09 USD
77463-898 77476-704 77486-756 77497-602
1-isobutyl-4-(methoxymethoxy)benzene ≥97%
1-isobutyl-4-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₁₂H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34779342
CAS Number: 2901083-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR