5-Bromo-2-(trifluoromethyl)pyridin-4-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
106823-1G
106823-500MG
106823-5G
106823-25G
77463-894EA
79.31
USD
77463-894
77476-700
77486-752
77497-594
5-Bromo-2-(trifluoromethyl)pyridin-4-ol ≥95%
5-Bromo-2-(trifluoromethyl)pyridin-4-ol
Precaución: For research use only
Formula:
C₆H₃BrF₃NO MW: 241.993 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13189951 CAS Number: 1196156-74-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...