3,5-Dichloro-2-(methylthio)isonicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

106223-1G 106223-500MG 106223-5G 106223-25G
77463-890EA 415.09 USD
77463-890 77476-696 77486-748 77497-588
3,5-Dichloro-2-(methylthio)isonicotinaldehyde ≥95%
3,5-Dichloro-2-(methylthio)isonicotinaldehyde

Precaución: For research use only

Formula: C₇H₅Cl₂NOS
MW: 222.092 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778958
CAS Number: 2918927-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR