2-Bromo-4-fluoro-5-isobutoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

106122-1G 106122-500MG 106122-5G 106122-25G
77463-886EA 393.46 USD
77463-886 77476-692 77486-744 77497-586
2-Bromo-4-fluoro-5-isobutoxybenzaldehyde ≥97%
2-Bromo-4-fluoro-5-isobutoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₂BrFO₂
MW: 275.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34778884
CAS Number: 2918881-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR