2,3,5-trimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

105467-1G 105467-500MG 105467-5G 105467-25G
77463-882EA 275.01 USD
77463-882 77476-688 77486-738 77497-566
2,3,5-trimethoxybenzaldehyde ≥95%
2,3,5-trimethoxybenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835365
CAS Number: 5556-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR