(2,4-Difluoro-6-methoxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

105145-1G 105145-500MG 105145-5G 105145-25G
77463-878EA 94.76 USD
77463-878 77476-686 77486-734 77497-554
(2,4-Difluoro-6-methoxyphenyl)boronic acid ≥95%
(2,4-Difluoro-6-methoxyphenyl)boronic acid

Precaución: For research use only

Formula: C₇H₇BF₂O₃
MW: 187.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414936
CAS Number: 1808997-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR