4-isopropoxy-3,5-diisopropylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

105079-1G 105079-500MG 105079-5G 105079-25G
77463-874EA 415.09 USD
77463-874 77476-682 77486-730 77497-546
4-isopropoxy-3,5-diisopropylbenzoic acid ≥95%
4-isopropoxy-3,5-diisopropylbenzoic acid

Precaución: For research use only

Formula: C₁₆H₂₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34778318
CAS Number: 2918851-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR