4-Bromo-2-chloro-6-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

104945-1G 104945-500MG 104945-5G
77463-870EA 1287.5 USD
77463-870 77476-678 77486-726
4-Bromo-2-chloro-6-methoxybenzaldehyde ≥95%
4-Bromo-2-chloro-6-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD28399041
CAS Number: 1936128-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR