2-(2,6-dibromo-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

104622-1G 104622-500MG 104622-5G 104622-25G
77463-866EA 415.09 USD
77463-866 77476-674 77486-722 77497-522
2-(2,6-dibromo-4-(trifluoromethyl)phenyl)-1,3-dioxolane ≥97%
2-(2,6-dibromo-4-(trifluoromethyl)phenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₇Br₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34778139
CAS Number: 2918841-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR