2-Bromo-1-ethoxy-4-propylbenzene ≥95%

Supplier: AOBChem USA

Warning

104530-1G 104530-500MG 104530-5G 104530-25G
77463-862EA 415.09 USD
77463-862 77476-670 77486-718 77497-518
2-Bromo-1-ethoxy-4-propylbenzene ≥95%
2-Bromo-1-ethoxy-4-propylbenzene

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34820598
CAS Number: 2918959-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR