1-(4-Chloro-2-methoxyphenyl)-4,4-dimethylcyclohexanol ≥95%

Supplier: AOBChem USA

Warning

104198-1G 104198-500MG 104198-5G 104198-25G
77463-854EA 415.09 USD
77463-854 77476-662 77486-710 77497-508
1-(4-Chloro-2-methoxyphenyl)-4,4-dimethylcyclohexanol ≥95%
1-(4-Chloro-2-methoxyphenyl)-4,4-dimethylcyclohexanol

Precaución: For research use only

Formula: C₁₅H₂₁ClO₂
MW: 268.779 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777961
CAS Number: 2918931-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR