4-(5-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

104081-1G 104081-500MG 104081-5G
77463-850EA 546.93 USD
77463-850 77476-658 77486-706
4-(5-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Fluoro-2-methoxyphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₅FO₃
MW: 226.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24305424
CAS Number: 1545811-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR