2-(6-Bromo-2,3-dimethoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

104032-1G 104032-500MG 104032-5G 104032-25G
77463-846EA 415.09 USD
77463-846 77476-654 77486-702 77497-502
2-(6-Bromo-2,3-dimethoxyphenyl)-1,3-dioxolane ≥97%
2-(6-Bromo-2,3-dimethoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₄
MW: 289.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34820483
CAS Number: 213698-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR