3,5-Dibromo-4-chlorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

103514-1G 103514-500MG 103514-5G 103514-25G
77463-842EA 415.09 USD
77463-842 77476-650 77486-698 77497-494
3,5-Dibromo-4-chlorobenzonitrile ≥97%
3,5-Dibromo-4-chlorobenzonitrile

Precaución: For research use only

Formula: C₇H₂Br₂ClN
Storage Temperature: Ambient
MDL Number: MFCD28669842
CAS Number: 1541255-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR