1-(2-Fluoro-5-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103414-1G 103414-500MG 103414-5G
77463-838EA 447.02 USD
77463-838 77476-646 77486-694
1-(2-Fluoro-5-methylphenyl)cyclobutanol ≥95%
1-(2-Fluoro-5-methylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₃FO
Storage Temperature: Ambient
MDL Number: MFCD21788695
CAS Number: 1526006-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR