1-(2,3-Difluoro-4-methylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

103379-1G 103379-500MG 103379-5G 103379-25G
77463-834EA 369.77 USD
77463-834 77476-642 77486-690 77497-486
1-(2,3-Difluoro-4-methylphenyl)cyclobutanol ≥95%
1-(2,3-Difluoro-4-methylphenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₁H₁₂F₂O
Storage Temperature: Ambient
MDL Number: MFCD30318454
CAS Number: 1693640-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR