4-(5-Bromo-2-fluoro-3-(methylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

103364-1G 103364-500MG 103364-5G 103364-25G
77463-830EA 415.09 USD
77463-830 77476-638 77486-686 77497-484
4-(5-Bromo-2-fluoro-3-(methylthio)phenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Bromo-2-fluoro-3-(methylthio)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₂H₁₄BrFO₂S
MW: 321.206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777547
CAS Number: 2918947-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR