1,2-Dibromo-3-fluoro-4-propoxybenzene ≥95%

Supplier: AOBChem USA

Warning

103296-1G 103296-500MG 103296-5G 103296-25G
77463-826EA 415.09 USD
77463-826 77476-634 77486-682 77497-480
1,2-Dibromo-3-fluoro-4-propoxybenzene ≥95%
1,2-Dibromo-3-fluoro-4-propoxybenzene

Precaución: For research use only

Formula: C₉H₉Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD34820268
CAS Number: 2969330-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR