4-(2-Bromo-3-fluoropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

102705-1G 102705-5G 102705-500MG 102705-25G
77463-818EA 415.09 USD
77463-818 77486-674 77476-626 77497-448
4-(2-Bromo-3-fluoropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥97%
4-(2-Bromo-3-fluoropyridin-4-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₀H₁₁BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD34777283
CAS Number: 2901095-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR