4-(5-Chlorothiophen-2-yl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

102691-1G 102691-5G 102691-500MG 102691-25G
77463-814EA 415.09 USD
77463-814 77486-670 77476-622 77497-444
4-(5-Chlorothiophen-2-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Chlorothiophen-2-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₉H₁₁ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD23865083
CAS Number: 1522784-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR