4,4-Dimethyl-1-(5-methylthiophen-2-yl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

102687-1G 102687-5G 102687-500MG
77463-810EA 546.93 USD
77463-810 77486-666 77476-618
4,4-Dimethyl-1-(5-methylthiophen-2-yl)cyclohexanol ≥95%
4,4-Dimethyl-1-(5-methylthiophen-2-yl)cyclohexanol

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21274533
CAS Number: 1486459-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR