4-(3-Chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

102583-1G 102583-5G 102583-500MG 102583-25G
77463-802EA 415.09 USD
77463-802 77486-658 77476-610 77497-426
4-(3-Chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥97%
4-(3-Chloropyridin-4-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₀H₁₂ClNO₂
MW: 213.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34777170
CAS Number: 2918851-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR