1-Cyclopentyl-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

102561-1G 102561-5G 102561-500MG
77463-798EA 622.12 USD
77463-798 77486-654 77476-606
1-Cyclopentyl-4-fluorobenzene ≥97%
1-Cyclopentyl-4-fluorobenzene

Precaución: For research use only

Formula: C₁₁H₁₃F
Storage Temperature: Ambient
MDL Number: MFCD15527414
CAS Number: 807335-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR