2-Bromo-5-(dimethoxymethyl)-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

102545-1G 102545-5G 102545-500MG 102545-25G
77463-794EA 415.09 USD
77463-794 77486-650 77476-602 77497-420
2-Bromo-5-(dimethoxymethyl)-3-methylpyridine ≥97%
2-Bromo-5-(dimethoxymethyl)-3-methylpyridine

Precaución: For research use only

Formula: C₉H₁₂BrNO₂
MW: 246.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28675338
CAS Number: 1131795-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR