1-(3-Bromophenyl)pentan-1-ol ≥97%

Supplier: AOBChem USA

Warning

102541-1G 102541-5G 102541-500MG
77463-790EA 487.19 USD
77463-790 77486-646 77476-598
1-(3-Bromophenyl)pentan-1-ol ≥97%
1-(3-Bromophenyl)pentan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16820772
CAS Number: 158918-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR