6-Bromo-2-(1,3-dioxolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

10210-1G 10210-5G 10210-500MG 10210-25G
77463-786EA 111.24 USD
77463-786 77486-642 77476-594 77497-394
6-Bromo-2-(1,3-dioxolan-2-yl)pyridine ≥97%
6-Bromo-2-(1,3-dioxolan-2-yl)pyridine

Precaución: For research use only

Formula: C₈H₈BrNO₂
MW: 230.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06209635
CAS Number: 34199-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR