4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

10152-1G 10152-5G 10152-500MG 10152-25G
77463-782EA 114.33 USD
77463-782 77486-638 77476-590 77497-380
4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid pinacol ester ≥97%
4-Benzyloxy-2-dimethylamino-pyrimidine-5-boronic acid pinacol ester

Precaución: For research use only

Formula: C₁₉H₂₆BN₃O₃
MW: 355.239 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08689617
CAS Number: 2121513-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR