(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

101220-1G 101220-5G 101220-500MG 101220-25G
77463-774EA 415.09 USD
77463-774 77486-630 77476-582 77497-364
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(6-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₅BrF₄S
MW: 289.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34776474
CAS Number: 2169262-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR