7-Methoxy-2-naphthaldehyde ≥95%

Supplier: AOBChem USA

Warning

100977-1G 100977-5G 100977-500MG
77463-770EA 659.2 USD
77463-770 77486-626 77476-578
7-Methoxy-2-naphthaldehyde ≥95%
7-Methoxy-2-naphthaldehyde

Precaución: For research use only

Formula: C₁₂H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD18412883
CAS Number: 5665-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR