(7-Methoxynaphthalen-2-yl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

100972-1G 100972-5G 100972-500MG
77463-766EA 515 USD
77463-766 77486-622 77476-574
(7-Methoxynaphthalen-2-yl)boronic acid ≥95%
(7-Methoxynaphthalen-2-yl)boronic acid

Precaución: For research use only

Formula: C₁₁H₁₁BO₃
MW: 202.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18412868
CAS Number: 313947-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR