Ethyl propionate ≥99.0%

Supplier: TCI America
Danger

Synonyms: Propionic acid ethyl ester

P0505-500ML P0505-25ML
TCP0505-500MLEA 43 USD
TCP0505-500ML TCP0505-25ML
Ethyl propionate ≥99.0%
Ethyl propionate

CAS Number: 105-37-3
MDL Number: MFCD00009308
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 9
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Propionic Acid Ethyl Ester

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 99 °C
Melting Pt: -74 °C
Density: 0.89 (20 °C)
Flash Pt: 9 °C
Storage Temperature: Ambient
MDL Number: MFCD00009308
CAS Number: 105-37-3
EINECS: 203-291-4
UN: 1195
ADR: 3,II
Merck Index: 13,03878
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95e2fe73279e9dde51a839b9e4b3e7cfa027f09b&vid=95e2fe73279e9dde51a839b9e4b3e7cfa027f09b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR