Pyrazine ≥98.0%
Supplier: TCI America
P0544-500G
P0544-25G
TCP0544-500GEA
241
USD
TCP0544-500G
TCP0544-25G
Pyrazine ≥98.0%
Pyrazine
CAS Number: 290-37-9
MDL Number: MFCD00006122
Molecular Formula: C4H4N2
Molecular Weight: 80.09
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 54
Flash Point (°C): 55
Formula:
C₄H₄N₂ MW: 80.09 g/mol Boiling Pt: 118 °C Melting Pt: 54 °C Density: 1.0311 g/cm³ (60.9 °C) Flash Pt: 55 °C |
MDL Number:
MFCD00006122 CAS Number: 290-37-9 EINECS: 206-027-6 UN: 1325 ADR: 4.1,III Merck Index: 13,08047 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...