Pinacyanol iodide 90,0
Supplier: TCI America
Synonyms:
Iodide, Quinaldine Blue, 1,1'-Diethyl-2,2'-carbocyanine Iodide
P0436-1G
P0436-5G
TCP0436-001GEA
114
USD
TCP0436-001G
TCP0436-005G
Pinacyanol iodide 90,0
Pinacyanol iodide
CAS Number: 605-91-4
MDL Number: MFCD00011975
Molecular Formula: C25H25IN2
Molecular Weight: 480.39
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Product Categories: Materials Chemistry, Cyanine & Squarylium Dyes (Functional Organic Dyes), Bioscience, Stains & Dyes, Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 1,1'-Diethyl-2,2'-carbocyanine Iodide, Quinaldine Blue, Iodide
Formula:
C₂₅H₂₅IN₂ MW: 480.39 g/mol |
MDL Number:
MFCD00011975 CAS Number: 605-91-4 |
|
Specification Test Results
Purity/Analysis Method | >90.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...